Assay Details
Target Gene Details
Entrez Gene ID: | 6364 |
Gene Name: | C-C motif chemokine ligand 20 |
Gene Aliases: |
CKb4, Exodus, LARC, MIP-3-alpha, MIP-3a, MIP3A, SCYA20, ST38 |
Location: |
Chr.2:227813842-227817564 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL20 | NM_001130046.1 | 4 | 412 | NP_001123518.1 |
| NM_004591.2 | 4 | 415 | NP_004582.1 | |
| BC020698.1 | 4 | 412 | AAH20698.1 | |
| D86955.1 | 4 | 403 | BAA13191.1 | |
| U64197.1 | 4 | 384 | AAB61534.1 | |
| U77035.1 | 4 | 345 | AAC50943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10221 | Chr.2:227715193 - 228041264 on Build GRCh38 | Loss |
|
| nsv834562 | Chr.2:227752511 - 227939244 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map