Assay Details
Target Gene Details
Entrez Gene ID: | 8828 |
Gene Name: | neuropilin 2 |
Gene Aliases: |
NP2, NPN2, PRO2714, VEGF165R2 |
Location: |
Chr.2:205682450-205798133 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRP2 | NM_003872.2 | 1 | 825 | NP_003863.2 |
| NM_018534.3 | 1 | 825 | NP_061004.3 | |
| NM_201264.1 | 1 | 825 | NP_957716.1 | |
| NM_201266.1 | 1 | 825 | NP_957718.1 | |
| NM_201267.1 | 1 | 825 | NP_957719.1 | |
| NM_201279.1 | 1 | 825 | NP_958436.1 | |
| XM_005246933.3 | 2 | 690 | XP_005246990.2 | |
| XM_005246934.3 | 2 | 363 | XP_005246991.2 | |
| XM_017005185.1 | 2 | 689 | XP_016860674.1 | |
| XM_017005186.1 | 2 | 690 | XP_016860675.1 | |
| XM_017005187.1 | 1 | 875 | XP_016860676.1 | |
| XM_017005188.1 | 1 | 874 | XP_016860677.1 | |
| AF016098.1 | AAC12922.1 | |||
| AF022859.1 | AAC51788.1 | |||
| AF022860.1 | AAC51789.1 | |||
| AF280544.1 | AAG41403.1 | |||
| AF280545.1 | AAG41404.1 | |||
| AF280546.1 | AAG41405.1 | |||
| AK130198.1 | 2 | 811 | ||
| AK290934.1 | 2 | 505 | ||
| AL833606.1 | 1 | 825 | ||
| BC009222.2 | 3 | 503 | AAH09222.1 | |
| BC101525.1 | AAI01526.1 | |||
| BC104770.1 | AAI04771.1 | |||
| BC117413.1 | ||||
| BC143238.1 | ||||
| BC143608.1 | ||||
| BX537423.1 | 1 | 826 | CAD97665.1 | |
| KJ534899.1 | ||||
| KJ534900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834514 | Chr.2:205575978 - 205738443 on Build GRCh38 | Gain |
|
| nsv3123 | Chr.2:205681544 - 205714569 on Build GRCh38 | Insertion |
|
| esv25775 | Chr.2:205407368 - 205806595 on Build GRCh38 | Gain |
|
| nsv584246 | Chr.2:205627674 - 205747861 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map