Assay Details
Target Gene Details
Entrez Gene ID: | 860 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA |
Location: |
Chr.6:45328142-45664032 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNX2 | NM_001015051.3 | NP_001015051.3 | ||
| NM_001024630.3 | NP_001019801.3 | |||
| XM_011514960.2 | XP_011513262.1 | |||
| XM_011514961.2 | XP_011513263.1 | |||
| XM_011514962.2 | XP_011513264.1 | |||
| XM_011514963.2 | XP_011513265.1 | |||
| XM_011514964.2 | XP_011513266.1 | |||
| XM_011514965.2 | XP_011513267.1 | |||
| XM_017011391.1 | XP_016866880.1 | |||
| XM_017011393.1 | XP_016866882.1 | |||
| XM_017011394.1 | XP_016866883.1 | |||
| XM_017011395.1 | XP_016866884.1 |
Target Gene Details
Entrez Gene ID: | 8464 |
Gene Name: | SPT3 homolog, SAGA and STAGA complex component |
Gene Aliases: |
SPT3, SPT3L |
Location: |
Chr.6:44808844-45378051 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT3H | NM_001261823.1 | NP_001248752.1 | ||
| NM_003599.3 | NP_003590.1 | |||
| NM_181356.2 | NP_852001.1 | |||
| XM_011514950.1 | XP_011513252.1 | |||
| AF073930.1 | AAC36098.1 | |||
| AK303838.1 | ||||
| AK313874.1 | ||||
| BC035816.1 | AAH35816.1 | |||
| BC050384.1 | AAH50384.1 | |||
| DA012115.1 |
More Information
Set Membership: |
|

Genomic Map