Assay Details
Target Gene Details
Entrez Gene ID: | 51552 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
FBP, RAB-14 |
Location: |
Chr.9:121178137-121202087 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB14 | NM_016322.3 | 8 | 987 | NP_057406.2 |
| AF112206.1 | 8 | 595 | AAF17194.1 | |
| AF152463.1 | 7 | 531 | AAF00150.1 | |
| AF203689.1 | 7 | 537 | AAF19400.1 | |
| AF498949.1 | 7 | 531 | AAM21097.1 | |
| AK023524.1 | 8 | 769 | BAB14598.1 | |
| AK090889.1 | 8 | 786 | ||
| AL162081.1 | 8 | 714 | CAB82414.1 | |
| AY553875.1 | 7 | 531 | AAS64573.1 | |
| BC006081.1 | 8 | 758 | AAH06081.1 | |
| CR457394.1 | 7 | 531 | CAG33675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950937 | Chr.9:121178823 - 121200622 on Build GRCh38 | Duplication |
|
| nsv820290 | Chr.9:121181543 - 121182897 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map