Assay Details
Target Gene Details
Entrez Gene ID: | 80781 |
Gene Name: | collagen type XVIII alpha 1 chain |
Gene Aliases: |
KNO, KNO1, KS |
Location: |
Chr.21:45405137-45513720 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL18A1 | NM_130445.3 | NP_569712.2 | ||
| AF018082.1 | AAC39659.1 | |||
| AK130835.1 | ||||
| BC063833.1 | AAH63833.1 | |||
| CN389577.1 |
Target Gene Details
Entrez Gene ID: | 378832 |
Gene Name: | COL18A1 antisense RNA 1 |
Gene Aliases: |
C21orf123, NCRNA00175, PRED80 |
Location: |
Chr.21:45419716-45425070 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL18A1-AS1 | NR_027498.1 | 3 | 1065 | |
| NR_028082.1 | 3 | 2149 | ||
| AK096071.1 | 3 | 1065 | BAC04694.1 | |
| BC111539.2 | 3 | 2149 | AAI11540.1 | |
| BC111540.2 | 3 | 1035 | AAI11541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516407 | Chr.21:45410958 - 45620742 on Build GRCh38 | Loss |
|
| esv2762114 | Chr.21:45260287 - 45647830 on Build GRCh38 | Gain |
|
| nsv1057607 | Chr.21:45333418 - 45463244 on Build GRCh38 | Gain |
|
| nsv953653 | Chr.21:45409686 - 45560386 on Build GRCh38 | Deletion |
|
| dgv4436n100 | Chr.21:45402338 - 45556768 on Build GRCh38 | Gain |
|
| dgv7900n54 | Chr.21:45300095 - 45428674 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115136569] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map