Assay Details
Target Gene Details
Entrez Gene ID: | 57062 |
Gene Name: | DEAD-box helicase 24 |
Gene Aliases: |
- |
Location: |
Chr.14:94050922-94081212 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX24 | NM_020414.3 | 7 | 2185 | NP_065147.1 |
| AB209382.1 | 5 | 4742 | BAD92619.1 | |
| AF134475.1 | 5 | 1001 | AAP97252.1 | |
| AF145022.1 | 5 | 1001 | AAP97274.1 | |
| AF214731.1 | 7 | 2184 | AAG02169.1 | |
| AK025162.1 | 7 | 2181 | BAB15079.1 | |
| AK294613.1 | 5 | 1023 | ||
| AK296726.1 | 8 | 1896 | ||
| AK297232.1 | 7 | 2049 | ||
| AK304504.1 | 6 | 1452 | ||
| AL136886.1 | 7 | 2161 | CAB66820.1 | |
| BC008847.2 | 7 | 2161 | AAH08847.1 | |
| BC009406.2 | 7 | 2161 | AAH09406.1 | |
| BC096826.1 | 8 | 1991 | AAH96826.1 | |
| BX537533.1 | 6 | 4809 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581391 | Chr.14:94035248 - 94057787 on Build GRCh38 | Loss |
|
| esv34067 | Chr.14:94024355 - 94160851 on Build GRCh38 | Loss |
|
| esv3635373 | Chr.14:94040151 - 94173989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map