Assay Details
Target Gene Details
Entrez Gene ID: | 51001 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
CGI-12, MTERFD1 |
Location: |
Chr.8:96239398-96261613 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTERF3 | NM_001286643.1 | 7 | 1123 | NP_001273572.1 |
| NM_015942.4 | 7 | 1123 | NP_057026.3 | |
| XM_006716570.3 | 7 | 1081 | XP_006716633.1 | |
| XM_011517054.2 | 6 | 790 | XP_011515356.1 | |
| AF132946.1 | 6 | 845 | AAD27721.1 | |
| AK001801.1 | 5 | 827 | ||
| AK309002.1 | 7 | 1120 | ||
| BC012995.2 | 7 | 1078 | AAH12995.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6318 | Chr.8:96219001 - 96245773 on Build GRCh38 | Insertion |
|
| nsv1020494 | Chr.8:96188447 - 96286981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map