Assay Details
Target Gene Details
Entrez Gene ID: | 93034 |
Gene Name: | 5'-nucleotidase, cytosolic IB |
Gene Aliases: |
AIRP, CN-IB, CN1B |
Location: |
Chr.2:18562871-18589580 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C1B | NM_001002006.2 | NP_001002006.1 | ||
| NM_001199086.1 | NP_001186015.1 | |||
| NM_001199087.1 | NP_001186016.1 | |||
| NM_001199088.1 | NP_001186017.1 | |||
| NM_033253.3 | NP_150278.2 | |||
| AF352326.1 | AAK83285.1 | |||
| AF356185.1 | AAK39108.1 | |||
| AF417165.1 | AAL11910.1 | |||
| AJ295254.1 | CAC44363.1 | |||
| AK093234.1 | BAC04103.1 | |||
| AK093464.1 | BAC04174.1 | |||
| AK097422.1 | ||||
| AK302084.1 | ||||
| AK302200.1 | ||||
| AK302795.1 | ||||
| BC136387.1 | ||||
| BC171761.1 |
Target Gene Details
Entrez Gene ID: | 100526794 |
Gene Name: | NT5C1B-RDH14 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:18554723-18589580 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C1B-RDH14 | NM_001199103.1 | NP_001186032.1 | ||
| NM_001199104.1 | NP_001186033.1 | |||
| AK057444.1 | BAB71489.1 | |||
| AK307653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589967 | Chr.2:18572204 - 18580279 on Build GRCh38 | Loss |
|
| esv2751837 | Chr.2:18240747 - 18831848 on Build GRCh38 | Gain |
|
| nsv581114 | Chr.2:18200727 - 18615490 on Build GRCh38 | Gain |
|
| nsv519085 | Chr.2:18574386 - 18709413 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map