Assay Details
Target Gene Details
Entrez Gene ID: | 4172 |
Gene Name: | minichromosome maintenance complex component 3 |
Gene Aliases: |
HCC5, P1-MCM3, P1.h, RLFB |
Location: |
Chr.6:52264014-52284881 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM3 | NM_001270472.1 | NP_001257401.1 | ||
| NM_002388.4 | NP_002379.3 | |||
| AK222586.1 | BAD96306.1 | |||
| AK299566.1 | ||||
| AK300754.1 | ||||
| AK301704.1 | ||||
| AK312928.1 | ||||
| AY032603.1 | AAK56392.1 | |||
| BC001626.1 | AAH01626.1 | |||
| BC003509.1 | AAH03509.2 | |||
| D38073.1 | BAA07267.1 | |||
| KU178170.1 | ||||
| KU178171.1 | ||||
| X62153.2 | CAA44078.2 |
More Information
Set Membership: |
|

Genomic Map