Assay Details
Target Gene Details
Entrez Gene ID: | 6133 |
Gene Name: | ribosomal protein L9 |
Gene Aliases: |
L9, NPC-A-16 |
Location: |
Chr.4:39454124-39458948 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL9 | NM_000661.4 | NP_000652.2 | ||
| NM_001024921.3 | NP_001020092.1 | |||
| AK297178.1 | ||||
| AL039355.2 | ||||
| AL832047.1 | CAH10410.1 | |||
| AY320412.1 | AAP73811.1 | |||
| AY376242.1 | AAQ82909.1 | |||
| BC100285.1 | AAI00286.1 | |||
| BM841336.1 | ||||
| U09953.1 | AAB01040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010189 | Chr.4:39423149 - 39523510 on Build GRCh38 | Gain |
|
| nsv1010784 | Chr.4:39437476 - 39571758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map