Assay Details
Target Gene Details
Entrez Gene ID: | 79869 |
Gene Name: | cleavage and polyadenylation specific factor 7 |
Gene Aliases: |
CFIm59 |
Location: |
Chr.11:61402648-61429992 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF7 | NM_001136040.2 | 3 | 288 | NP_001129512.1 |
| NM_001142565.1 | 3 | 288 | NP_001136037.1 | |
| NM_024811.3 | 3 | 364 | NP_079087.3 | |
| XM_005274298.4 | 3 | 374 | XP_005274355.1 | |
| XM_005274299.4 | 3 | 291 | XP_005274356.1 | |
| XM_005274303.4 | 3 | 245 | XP_005274360.1 | |
| XM_011545257.2 | 3 | 374 | XP_011543559.1 | |
| XM_011545258.2 | 3 | 245 | XP_011543560.1 | |
| XM_011545259.2 | 3 | 245 | XP_011543561.1 | |
| XM_011545260.2 | 3 | 288 | XP_011543562.1 | |
| XM_011545261.2 | 3 | 289 | XP_011543563.1 | |
| XM_011545262.2 | 3 | 246 | XP_011543564.1 | |
| XM_011545263.2 | 3 | 245 | XP_011543565.1 | |
| XM_017018345.1 | 3 | 374 | XP_016873834.1 | |
| AJ275970.1 | 2 | 154 | CAC81661.1 | |
| AK092037.1 | 2 | 218 | ||
| AK096343.1 | 3 | 244 | ||
| AK294578.1 | 3 | 244 | ||
| AK307190.1 | 3 | 410 | ||
| AL832546.1 | ||||
| AL832667.1 | 3 | 279 | ||
| AL833332.1 | 3 | 244 | ||
| BC018135.1 | 3 | 254 | AAH18135.1 | |
| BX537888.1 | 3 | 269 | CAD97884.1 | |
| DA194038.1 | 3 | 364 | ||
| DA804579.1 | 3 | 288 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832178 | Chr.11:61402074 - 61573390 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map