Assay Details
Target Gene Details
Entrez Gene ID: | 3097 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 2 |
Gene Aliases: |
HIV-EP2, MBP-2, MIBP1, MRD43, SHN2, ZAS2, ZNF40B |
Location: |
Chr.6:142751467-142945201 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIVEP2 | NM_006734.3 | 10 | 7682 | NP_006725.3 |
| XM_017010805.1 | 10 | 7647 | XP_016866294.1 | |
| AK226163.1 | 6 | 5160 | ||
| AK296531.1 | 6 | 1930 | ||
| CR936782.1 | 10 | 7682 | ||
| DQ231041.1 | 6 | 6939 | ABB16331.1 | |
| X65644.1 | 6 | 7116 | CAA46596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830828 | Chr.6:142648997 - 142795380 on Build GRCh38 | Gain |
|
| nsv830829 | Chr.6:142737271 - 142896940 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map