Assay Details
Target Gene Details
Entrez Gene ID: | 7873 |
Gene Name: | mesencephalic astrocyte derived neurotrophic factor |
Gene Aliases: |
ARMET, ARP |
Location: |
Chr.3:51385237-51389397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MANF | NM_006010.5 | NP_006001.4 | ||
| AK292243.1 | ||||
| BC007282.1 | AAH07282.1 | |||
| BC113588.1 | ||||
| BC113590.1 | ||||
| BE514267.1 | ||||
| BT007110.1 | AAP35774.1 | |||
| CN410911.1 | ||||
| M83751.1 | AAB08753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590277 | Chr.3:51370620 - 51466575 on Build GRCh38 | Loss |
|
| dgv8321n54 | Chr.3:51355615 - 51395464 on Build GRCh38 | Loss |
|
| nsv590276 | Chr.3:51358140 - 51431657 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map