Assay Details
Target Gene Details
Entrez Gene ID: | 57447 |
Gene Name: | NDRG family member 2 |
Gene Aliases: |
SYLD |
Location: |
Chr.14:21016763-21070872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG2 | NM_001282211.1 | NP_001269140.1 | ||
| AK293514.1 |
Target Gene Details
Entrez Gene ID: | 440163 |
Gene Name: | ribonuclease A family member 13 (inactive) |
Gene Aliases: |
HEL-S-86p, RAL1 |
Location: |
Chr.14:21032820-21034785 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE13 | NM_001012264.3 | 2 | 778 | NP_001012264.1 |
| BC044831.1 | 2 | 778 | AAH44831.1 | |
| FJ237363.1 | 2 | 778 |
Target Gene Details
Entrez Gene ID: | 122664 |
Gene Name: | tubulin polymerization promoting protein family member 2 |
Gene Aliases: |
C14orf8, CT152, P18, p25beta |
Location: |
Chr.14:21024262-21036352 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPPP2 | XM_011536416.1 | XP_011534718.1 | ||
| XM_011536420.2 | XP_011534722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
| nsv1039369 | Chr.14:21030895 - 21062142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map