Assay Details
Target Gene Details
Entrez Gene ID: | 5423 |
Gene Name: | DNA polymerase beta |
Gene Aliases: |
- |
Location: |
Chr.8:42338452-42371813 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLB | NM_002690.2 | NP_002681.1 | ||
| XM_005273535.3 | XP_005273592.1 | |||
| XM_005273536.3 | XP_005273593.1 | |||
| XM_005273537.3 | XP_005273594.1 | |||
| XM_005273538.2 | XP_005273595.1 | |||
| XM_005273539.2 | XP_005273596.1 | |||
| XM_005273540.4 | XP_005273597.1 | |||
| XM_006716353.2 | XP_006716416.1 | |||
| XM_017013583.1 | XP_016869072.1 | |||
| XM_017013584.1 | XP_016869073.1 | |||
| AK223537.1 | BAD97257.1 | |||
| AK294025.1 | ||||
| AK314976.1 | ||||
| BC100288.1 | AAI00289.1 | |||
| BC106909.1 | AAI06910.1 | |||
| BP370570.1 | ||||
| CR536503.1 | CAG38741.1 | |||
| CR541802.1 | CAG46601.1 | |||
| D29013.1 | BAA06099.1 | |||
| JQ664683.1 | ||||
| JQ927442.1 | ||||
| L11607.1 | AAB59441.1 | |||
| M13140.1 | AAA60133.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6169 | Chr.8:42298991 - 42342579 on Build GRCh38 | Deletion |
|
| nsv831301 | Chr.8:42244080 - 42473666 on Build GRCh38 | Loss |
|
| nsv508508 | Chr.8:42256602 - 42355479 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map