Assay Details
Target Gene Details
Entrez Gene ID: | 3359 |
Gene Name: | 5-hydroxytryptamine receptor 3A |
Gene Aliases: |
5-HT-3, 5-HT3A, 5-HT3R, 5HT3R, HTR3 |
Location: |
Chr.11:113975075-113990313 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3A | NM_000869.5 | NP_000860.2 | ||
| NM_001161772.2 | NP_001155244.1 | |||
| NM_213621.3 | NP_998786.2 | |||
| NR_046363.1 | ||||
| AF498984.1 | AAM21131.1 | |||
| AJ003078.1 | CAA05851.1 | |||
| AJ003079.1 | CAA05852.1 | |||
| AJ003080.1 | CAA05853.1 | |||
| AK299973.1 | ||||
| AK304630.1 | ||||
| AK313896.1 | ||||
| BC002354.2 | AAH02354.1 | |||
| BC004453.1 | AAH04453.2 | |||
| BT007204.1 | AAP35868.1 | |||
| CD014118.1 | ||||
| D49394.1 | BAA08387.1 | |||
| S82612.1 | AAB37533.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627783 | Chr.11:113969123 - 113990232 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map