Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD13 | NM_001243476.2 | 16 | 3445 | NP_001230405.1 |
| NM_052851.2 | 12 | 3035 | NP_443083.1 | |
| NM_178006.3 | 12 | 3098 | NP_821074.1 | |
| NM_178007.2 | 12 | 3005 | NP_821075.1 | |
| XM_011535299.2 | 15 | 3191 | XP_011533601.1 | |
| XM_017020834.1 | 12 | 3203 | XP_016876323.1 | |
| XM_017020835.1 | 14 | 3124 | XP_016876324.1 | |
| AK094704.1 | 11 | 1996 | ||
| AK095114.1 | 8 | 1718 | ||
| AK312888.1 | 12 | 3091 | ||
| AL049801.1 | 12 | 2991 | CAB42562.1 | |
| AY082589.1 | 12 | 3093 | AAL91648.1 | |
| AY082590.1 | 12 | 3017 | AAL91649.1 | |
| AY082591.1 | 12 | 2991 | AAL91650.1 | |
| AY366448.1 | 12 | 3033 | AAQ72791.1 | |
| BX641149.1 | CAE46065.1 | |||
| BX647695.1 | 16 | 3405 | CAI46026.1 |
More Information
Set Membership: |
|

Genomic Map