Assay Details
Target Gene Details
Entrez Gene ID: | 5605 |
Gene Name: | mitogen-activated protein kinase kinase 2 |
Gene Aliases: |
CFC4, MAPKK2, MEK2, MKK2, PRKMK2 |
Location: |
Chr.19:4090321-4124184 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP2K2 | NM_030662.3 | NP_109587.1 | ||
| XM_006722799.2 | XP_006722862.1 | |||
| XM_017026989.1 | XP_016882478.1 | |||
| XM_017026990.1 | XP_016882479.1 | |||
| AK093125.1 | ||||
| AK126446.1 | ||||
| BC000471.2 | AAH00471.1 | |||
| BC018645.2 | AAH18645.1 | |||
| BX538081.1 | CAD98005.1 | |||
| L11285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470115 | Chr.19:4024022 - 4098778 on Build GRCh38 | Loss |
|
| nsv578435 | Chr.19:4011267 - 4098778 on Build GRCh38 | Loss |
|
| esv2717979 | Chr.19:4005406 - 4159344 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146376140] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map