Assay Details
Target Gene Details
Entrez Gene ID: | 353299 |
Gene Name: | regulator of G-protein signaling like 1 |
Gene Aliases: |
RGSL, RGSL2 |
Location: |
Chr.1:182447877-182560597 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGSL1 | NM_001137669.1 | NP_001131141.1 | ||
| XM_005245137.1 | XP_005245194.1 | |||
| XM_011509494.2 | XP_011507796.1 | |||
| XM_011509496.1 | XP_011507798.1 | |||
| XM_011509500.1 | XP_011507802.1 | |||
| XM_011509503.1 | XP_011507805.1 | |||
| XM_011509504.2 | XP_011507806.1 | |||
| XM_017001188.1 | XP_016856677.1 | |||
| XM_017001189.1 | XP_016856678.1 | |||
| XM_017001191.1 | XP_016856680.1 | |||
| XM_017001192.1 | XP_016856681.1 | |||
| XM_017001193.1 | XP_016856682.1 | |||
| XM_017001194.1 | XP_016856683.1 | |||
| AK302768.1 | ||||
| BC142944.1 | ||||
| DB097250.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map