Assay Details
Target Gene Details
Entrez Gene ID: | 29855 |
Gene Name: | ubinuclein 1 |
Gene Aliases: |
VT, VT4 |
Location: |
Chr.16:4846665-4882401 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBN1 | NM_001079514.2 | 2 | 776 | NP_001072982.1 |
| NM_001288656.1 | 1 | 210 | NP_001275585.1 | |
| XM_005255277.3 | 2 | 961 | XP_005255334.1 | |
| XM_005255278.3 | 2 | 1925 | XP_005255335.1 | |
| XM_005255279.3 | 2 | 961 | XP_005255336.1 | |
| XM_011522465.1 | 2 | 961 | XP_011520767.1 | |
| XM_011522467.1 | 2 | 961 | XP_011520769.1 | |
| XM_017023189.1 | 2 | 961 | XP_016878678.1 | |
| XM_017023190.1 | 2 | 961 | XP_016878679.1 | |
| XM_017023192.1 | 2 | 961 | XP_016878681.1 | |
| AF108460.1 | 2 | 271 | AAF31755.1 | |
| AF108461.1 | 1 | 860 | AAF31756.1 | |
| AK056282.1 | 2 | 1925 | ||
| AK300131.1 | 1 | 210 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26756 | Chr.16:4772529 - 4855231 on Build GRCh38 | Gain+Loss |
|
| esv2422516 | Chr.16:4775920 - 5160979 on Build GRCh38 | Deletion |
|
| nsv510674 | Chr.16:4832446 - 4882731 on Build GRCh38 | Deletion |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| esv3637707 | Chr.16:4823818 - 4874308 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map