Assay Details
Target Gene Details
Entrez Gene ID: | 84904 |
Gene Name: | Rho guanine nucleotide exchange factor 39 |
Gene Aliases: |
C9orf100 |
Location: |
Chr.9:35658875-35665281 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF39 | NM_032818.2 | NP_116207.2 | ||
| XM_011518057.2 | XP_011516359.1 | |||
| XM_017015225.1 | XP_016870714.1 | |||
| XM_017015226.1 | XP_016870715.1 | |||
| XM_017015227.1 | XP_016870716.1 | |||
| XM_017015228.1 | XP_016870717.1 | |||
| AK027548.1 | BAB55192.1 | |||
| AK225501.1 | ||||
| AK225529.1 | ||||
| AK303513.1 | ||||
| AY390225.1 | AAR01010.1 | |||
| AY390226.1 | AAR01011.1 | |||
| BC033666.1 | AAH33666.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466385 | Chr.9:35662254 - 35705089 on Build GRCh38 | Loss |
|
| esv3620382 | Chr.9:35662903 - 35664509 on Build GRCh38 | Loss |
|
| esv3544805 | Chr.9:35662936 - 35664587 on Build GRCh38 | Deletion |
|
| esv2665709 | Chr.9:35662937 - 35664534 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map