Assay Details
Target Gene Details
Entrez Gene ID: | 64506 |
Gene Name: | cytoplasmic polyadenylation element binding protein 1 |
Gene Aliases: |
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1 |
Location: |
Chr.15:82543201-82648016 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB1 | NM_001079533.1 | NP_001073001.1 | ||
| NM_001079534.1 | NP_001073002.1 | |||
| NM_001079535.1 | NP_001073003.1 | |||
| NM_001288819.1 | NP_001275748.1 | |||
| NM_001288820.1 | NP_001275749.1 | |||
| NM_030594.4 | NP_085097.3 | |||
| AF329402.1 | AAK01239.1 | |||
| AF329403.1 | AAK01240.1 | |||
| AK023265.1 | BAB14496.1 | |||
| AK294280.1 | ||||
| AK300105.1 | ||||
| AK301728.1 | ||||
| BC035348.1 | AAH35348.1 | |||
| BC050629.1 | AAH50629.1 | |||
| BX649159.1 | ||||
| DA340952.1 | ||||
| DA705313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483125 | Chr.15:82448179 - 82617091 on Build GRCh38 | Loss |
|
| nsv833071 | Chr.15:82454948 - 82641759 on Build GRCh38 | Gain |
|
| nsv1035899 | Chr.15:82439721 - 82646980 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map