Assay Details
Target Gene Details
Entrez Gene ID: | 5920 |
Gene Name: | retinoic acid receptor responder 3 |
Gene Aliases: |
HRASLS4, HRSL4, PLA1/2-3, RIG1, TIG3 |
Location: |
Chr.11:63536801-63546458 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RARRES3 | NM_004585.4 | 3 | 351 | NP_004576.2 |
| AB030815.1 | 3 | 307 | BAB08109.1 | |
| AB453252.1 | 2 | 283 | ||
| AF060228.1 | 3 | 312 | AAC84000.1 | |
| AF092922.1 | 3 | 344 | AAF02294.1 | |
| AK293357.1 | 3 | 336 | ||
| AK312110.1 | 3 | 336 | ||
| BC009678.1 | 3 | 331 | AAH09678.1 | |
| DA973023.1 | 3 | 353 | ||
| R55646.1 | 3 | 323 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555179 | Chr.11:63465891 - 63552767 on Build GRCh38 | Loss |
|
| nsv509417 | Chr.11:63485643 - 63569743 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map