Assay Details
Target Gene Details
Entrez Gene ID: | 4281 |
Gene Name: | midline 1 |
Gene Aliases: |
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY |
Location: |
Chr.X:10445310-11111264 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
More Information
Set Membership: |
|

Genomic Map