Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL1 | NM_001128150.1 | 4 | 1597 | NP_001121622.1 |
| NM_016467.4 | 5 | 1707 | NP_057551.1 | |
| XM_005246970.2 | 4 | 1626 | XP_005247027.1 | |
| XM_011512199.2 | XP_011510501.1 | |||
| XM_017005347.1 | 5 | 1746 | XP_016860836.1 | |
| XM_017005348.1 | XP_016860837.1 | |||
| AA603217.1 | 1 | 517 | ||
| AI674061.1 | ||||
| AK074756.1 | 4 | 1596 | BAC11184.1 | |
| AK075160.1 | 3 | 1598 | BAC11441.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115591984] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map