Assay Details
Target Gene Details
Entrez Gene ID: | 51225 |
Gene Name: | ABI family member 3 |
Gene Aliases: |
NESH, SSH3BP3 |
Location: |
Chr.17:49210227-49223225 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABI3 | NM_001135186.1 | 8 | 1849 | NP_001128658.1 |
| NM_016428.2 | 8 | 1867 | NP_057512.1 | |
| XM_005257429.2 | 8 | 1533 | XP_005257486.1 | |
| XM_011524873.1 | 8 | 1530 | XP_011523175.1 | |
| XM_017024721.1 | 8 | 1509 | XP_016880210.1 | |
| AA808681.1 | 1 | 267 | ||
| AB037886.1 | 8 | 1867 | BAA90667.1 | |
| AL136709.1 | CAB66644.1 | |||
| BC007780.2 | 8 | 1373 | AAH07780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953924 | Chr.17:49219339 - 49227238 on Build GRCh38 | Deletion |
|
| nsv2074 | Chr.17:49208171 - 49233994 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map