Assay Details
Target Gene Details
Entrez Gene ID: | 132724 |
Gene Name: | transmembrane protease, serine 11B |
Gene Aliases: |
- |
Location: |
Chr.4:68226653-68245694 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS11B | NM_182502.3 | 8 | 961 | NP_872308.2 |
| XM_011531608.2 | 8 | 961 | XP_011529910.1 | |
| XM_011531609.1 | XP_011529911.1 | |||
| AK290904.1 | 8 | 961 | ||
| AL833167.1 | 8 | 959 | CAD91168.1 | |
| BC126195.1 | 8 | 883 | ||
| BX537945.1 | 8 | 959 | CAD97913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520487 | Chr.4:68218235 - 68229479 on Build GRCh38 | Loss |
|
| nsv594506 | Chr.4:68183925 - 68357339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map