Assay Details
Target Gene Details
Entrez Gene ID: | 4885 |
Gene Name: | neuronal pentraxin 2 |
Gene Aliases: |
NARP, NP-II, NP2 |
Location: |
Chr.7:98617285-98629869 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPTX2 | NM_002523.2 | 3 | 878 | NP_002514.1 |
| AK311250.1 | 2 | 569 | ||
| BC009924.2 | 3 | 870 | AAH09924.1 | |
| BC034781.1 | 3 | 866 | AAH34781.1 | |
| BC035339.1 | 3 | 871 | AAH35339.1 | |
| BC048275.1 | 3 | 836 | AAH48275.2 | |
| U26662.1 | 3 | 594 | AAA92296.1 |
More Information
Set Membership: |
|

Genomic Map