Assay Details
Target Gene Details
Entrez Gene ID: | 79625 |
Gene Name: | neuron derived neurotrophic factor |
Gene Aliases: |
C4orf31, NORD |
Location: |
Chr.4:121035627-121072518 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDNF | NM_024574.3 | 4 | 1181 | NP_078850.3 |
| AK026844.1 | 1 | 266 | BAB15573.1 | |
| AK289615.1 | 4 | 1089 | ||
| AY358821.1 | 4 | 753 | AAQ89180.1 | |
| BC019351.1 | 4 | 1077 |
More Information
Set Membership: |
|

Genomic Map