Assay Details
Target Gene Details
Entrez Gene ID: | 1411 |
Gene Name: | crystallin beta A1 |
Gene Aliases: |
CRYB1, CTRCT10 |
Location: |
Chr.17:29246857-29254494 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYBA1 | NM_005208.4 | NP_005199.2 | ||
| XM_017024198.1 | XP_016879687.1 | |||
| BC069537.1 | AAH69537.1 | |||
| BC113693.1 | ||||
| KM230602.1 | ||||
| KM230603.1 | ||||
| KM230604.1 | ||||
| KM230605.1 | ||||
| KR336660.1 | ||||
| KR336661.1 | ||||
| U59058.1 | AAC50971.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72819448] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map