Assay Details
Target Gene Details
Entrez Gene ID: | 1786 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 1 |
Gene Aliases: |
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI |
Location: |
Chr.19:10133344-10195135 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNMT1 | NM_001130823.2 | 29 | 3196 | NP_001124295.1 |
| NM_001318730.1 | 28 | 3067 | NP_001305659.1 | |
| NM_001318731.1 | 29 | 3075 | NP_001305660.1 | |
| NM_001379.3 | 28 | 3148 | NP_001370.1 | |
| AB209413.2 | 28 | 2874 | BAD92650.1 | |
| AF180682.1 | 26 | 2655 | AAF23609.1 | |
| AK122759.1 | 1 | 833 | ||
| BC126227.1 | 29 | 3140 | ||
| BC144093.1 | 29 | 3140 | ||
| X63692.1 | 28 | 3149 | CAA45219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map