Assay Details
Target Gene Details
Entrez Gene ID: | 8395 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase type 1 beta |
Gene Aliases: |
MSS4, STM7 |
Location: |
Chr.9:68705217-69009176 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP5K1B | NM_001278253.1 | NP_001265182.1 | ||
| NM_003558.3 | NP_003549.1 | |||
| XM_005252261.3 | XP_005252318.1 | |||
| XM_006717301.1 | XP_006717364.1 | |||
| AK292734.1 | ||||
| AK295587.1 | ||||
| CB988216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026962 | Chr.9:68335100 - 68742689 on Build GRCh38 | Gain |
|
| esv2422258 | Chr.9:68619929 - 69048424 on Build GRCh38 | Duplication |
|
| esv3891677 | Chr.9:68431371 - 68730207 on Build GRCh38 | Gain |
|
| esv3302888 | Chr.9:68676237 - 68721189 on Build GRCh38 | Tandem Duplication |
|
| esv3620546 | Chr.9:68675237 - 68722866 on Build GRCh38 | Gain |
|
| esv2759693 | Chr.9:68676191 - 68738827 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map