Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Gene Details
Entrez Gene ID: | 26499 |
Gene Name: | pleckstrin 2 |
Gene Aliases: |
- |
Location: |
Chr.14:67386979-67412200 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEK2 | NM_016445.2 | 5 | 689 | NP_057529.1 |
| XM_011536644.2 | 5 | 663 | XP_011534946.1 | |
| AF228603.1 | 5 | 600 | AAF34791.1 | |
| BC001226.1 | 5 | 633 | AAH01226.1 | |
| BC008056.2 | 5 | 543 | AAH08056.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751285 | Chr.14:66898030 - 67420064 on Build GRCh38 | Gain |
|
| nsv564961 | Chr.14:67266144 - 67637063 on Build GRCh38 | Gain |
|
| nsv509539 | Chr.14:67383566 - 67405348 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map