Assay Details
Target Gene Details
Entrez Gene ID: | 25945 |
Gene Name: | nectin cell adhesion molecule 3 |
Gene Aliases: |
CD113, CDW113, NECTIN-3, PPR3, PRR3, PVRL3, PVRR3 |
Location: |
Chr.3:111071759-111201444 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECTIN3 | NM_001243286.1 | 2 | 612 | NP_001230215.1 |
| NM_001243288.1 | 2 | 536 | NP_001230217.1 | |
| NM_015480.2 | 2 | 612 | NP_056295.1 | |
| XM_011512663.1 | 3 | 676 | XP_011510965.1 | |
| XM_017006123.1 | 3 | 676 | XP_016861612.1 | |
| XM_017006124.1 | 3 | 333 | XP_016861613.1 | |
| XM_017006125.1 | 2 | 536 | XP_016861614.1 | |
| XM_017006126.1 | 2 | 607 | XP_016861615.1 | |
| XM_017006127.1 | 2 | 699 | XP_016861616.1 | |
| AF282874.1 | 2 | 353 | AAF97597.1 | |
| AK075105.1 | 2 | 536 | BAC11404.1 | |
| BC067808.1 | 2 | 495 | AAH67808.1 | |
| BI464010.1 | 2 | 623 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591275 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain |
|
| nsv591276 | Chr.3:111103649 - 111135349 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map