Assay Details
Target Gene Details
Entrez Gene ID: | 55616 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 |
Gene Aliases: |
ACAP4, CENTB6, DDEFL1, UPLC1 |
Location: |
Chr.1:23428563-23484631 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASAP3 | NM_001143778.1 | 18 | 1989 | NP_001137250.1 |
| NM_017707.3 | 19 | 2016 | NP_060177.2 | |
| XM_011541755.1 | 19 | 1956 | XP_011540057.1 | |
| XM_017001685.1 | 19 | 1950 | XP_016857174.1 | |
| XM_017001686.1 | 19 | 1956 | XP_016857175.1 | |
| XM_017001687.1 | 19 | 1923 | XP_016857176.1 | |
| XM_017001688.1 | 19 | 1912 | XP_016857177.1 | |
| XM_017001689.1 | 19 | 1973 | XP_016857178.1 | |
| AB056749.1 | 19 | 1937 | BAB85677.1 | |
| AK000206.1 | 4 | 440 | BAA91008.1 | |
| AK000253.1 | 9 | 930 | BAA91033.1 | |
| AK074319.1 | 3 | 299 | ||
| AK092291.1 | 19 | 1938 | ||
| AK295100.1 | 18 | 1911 | ||
| AK299359.1 | 19 | 1912 | ||
| BC023519.2 | AAH23519.1 | |||
| BC052305.1 | 5 | 456 | AAH52305.1 | |
| BC060786.1 | 19 | 1964 | AAH60786.1 |
More Information
Set Membership: |
|

Genomic Map