Assay Details
Target Gene Details
Entrez Gene ID: | 23557 |
Gene Name: | SNAP associated protein |
Gene Aliases: |
BLOC1S7, BLOS7, BORCS3, SNAPAP |
Location: |
Chr.1:153658654-153661852 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAPIN | NM_012437.5 | NP_036569.1 | ||
| NR_052019.1 | ||||
| NR_052020.1 | ||||
| AF086837.1 | AAD11417.1 | |||
| AK024555.1 | BAB14927.1 | |||
| BC000761.2 | AAH00761.1 | |||
| BC004494.2 | AAH04494.1 | |||
| BE278772.1 | ||||
| BM051828.1 | ||||
| BT006753.1 | AAP35399.1 | |||
| CD671119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464028 | Chr.1:153649744 - 153750561 on Build GRCh38 | Gain |
|
| dgv582n54 | Chr.1:153639657 - 153750561 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map