Assay Details
Target Gene Details
Entrez Gene ID: | 373509 |
Gene Name: | ubiquitin specific peptidase 50 |
Gene Aliases: |
- |
Location: |
Chr.15:50500562-50546705 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP50 | NM_203494.4 | 7 | 1214 | NP_987090.2 |
| AJ583818.2 | CAE47745.2 | |||
| DB513975.2 | 4 | 537 |
Target Gene Details
Entrez Gene ID: | 9101 |
Gene Name: | ubiquitin specific peptidase 8 |
Gene Aliases: |
HumORF8, SPG59, UBPY |
Location: |
Chr.15:50424359-50501083 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP8 | NM_001128610.2 | 20 | 5352 | NP_001122082.1 |
| NM_001283049.1 | 17 | 4894 | NP_001269978.1 | |
| NM_005154.4 | 20 | 5212 | NP_005145.3 | |
| XM_006720761.3 | 21 | 5532 | XP_006720824.1 | |
| AA399952.1 | 1 | 345 | ||
| AA773713.1 | ||||
| AI276208.1 | 1 | 357 | ||
| CA312140.1 | 1 | 358 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv317n27 | Chr.15:50478671 - 50897510 on Build GRCh38 | Gain |
|
| nsv524613 | Chr.15:50364252 - 50576582 on Build GRCh38 | Loss |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| nsv569398 | Chr.15:50496928 - 51002798 on Build GRCh38 | Loss |
|
| dgv2594n100 | Chr.15:50469213 - 50823338 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map