Assay Details
Target Gene Details
Entrez Gene ID: | 254956 |
Gene Name: | MORN repeat containing 5 |
Gene Aliases: |
C9orf113, C9orf18 |
Location: |
Chr.9:122159661-122200088 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MORN5 | XM_006717034.3 | XP_006717097.1 | ||
| XM_017014578.1 | 1 | 153 | XP_016870067.1 |
Target Gene Details
Entrez Gene ID: | 4702 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A8 |
Gene Aliases: |
CI-19KD, CI-PGIV, PGIV |
Location: |
Chr.9:122132466-122159819 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA8 | NM_001318195.1 | NP_001305124.1 | ||
| NM_014222.2 | NP_055037.1 | |||
| AK301520.1 | ||||
| BU602971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6702 | Chr.9:122151820 - 122184094 on Build GRCh38 | Insertion |
|
| nsv831715 | Chr.9:122066612 - 122226720 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map