Assay Details
Target Gene Details
Entrez Gene ID: | 9860 |
Gene Name: | leucine rich repeats and immunoglobulin like domains 2 |
Gene Aliases: |
LIG-2, LIG2, UFS2 |
Location: |
Chr.1:113073170-113132260 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIG2 | NM_001312686.1 | NP_001299615.1 | ||
| NM_014813.2 | NP_055628.1 | |||
| XM_005271369.2 | XP_005271426.1 | |||
| XM_017002952.1 | XP_016858441.1 | |||
| AB018349.1 | ||||
| AK291031.1 | ||||
| BC117368.1 | ||||
| BC117370.1 | ||||
| CR936669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437002 | Chr.1:113049966 - 113116265 on Build GRCh38 | Loss |
|
| esv3587231 | Chr.1:113087524 - 113209293 on Build GRCh38 | Loss |
|
| nsv999915 | Chr.1:112817523 - 113142211 on Build GRCh38 | Gain |
|
| esv24626 | Chr.1:112832815 - 113310193 on Build GRCh38 | Loss |
|
| esv3894045 | Chr.1:112927499 - 113179081 on Build GRCh38 | Gain |
|
| esv3539690 | Chr.1:113090379 - 113113110 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map