Assay Details
Target Gene Details
Entrez Gene ID: | 4753 |
Gene Name: | neural EGFL like 2 |
Gene Aliases: |
NRP2 |
Location: |
Chr.12:44508275-44921848 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELL2 | NM_001145107.1 | 21 | 2714 | NP_001138579.1 |
| NM_001145108.1 | 20 | 3034 | NP_001138580.1 | |
| NM_001145109.1 | 20 | 2539 | NP_001138581.1 | |
| NM_001145110.1 | 21 | 2600 | NP_001138582.1 | |
| NM_006159.2 | 21 | 2641 | NP_006150.1 | |
| XM_005268905.3 | 21 | 2794 | XP_005268962.1 | |
| XM_011538396.1 | 22 | 2590 | XP_011536698.1 | |
| XM_017019341.1 | 21 | 2970 | XP_016874830.1 | |
| XM_017019342.1 | 21 | 2653 | XP_016874831.1 | |
| XM_017019343.1 | 21 | 2809 | XP_016874832.1 | |
| XM_017019344.1 | 22 | 2605 | XP_016874833.1 | |
| AK055385.1 | 1 | 1770 | ||
| AK095637.1 | 21 | 2868 | ||
| AK295125.1 | 21 | 2600 | ||
| AK299277.1 | 20 | 2495 | ||
| AK299733.1 | 21 | 2714 | ||
| AK309825.1 | 21 | 2484 | ||
| AK315960.1 | 20 | 2468 | ||
| AK316058.1 | 21 | 2961 | ||
| BC020544.1 | 20 | 2741 | AAH20544.1 | |
| D83018.1 | 21 | 2535 | BAA11681.1 | |
| D89629.1 | BAB46925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049818 | Chr.12:44092509 - 44740000 on Build GRCh38 | Gain |
|
| nsv1051285 | Chr.12:44054046 - 45582132 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map