Assay Details
Target Gene Details
Entrez Gene ID: | 1015 |
Gene Name: | cadherin 17 |
Gene Aliases: |
CDH16, HPT-1, HPT1 |
Location: |
Chr.8:94127162-94217303 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH17 | NM_001144663.1 | 18 | 3506 | NP_001138135.1 |
| NM_004063.3 | 18 | 3505 | NP_004054.3 | |
| XM_011516790.1 | 17 | 3264 | XP_011515092.1 | |
| AI566493.1 | 1 | 198 | ||
| AK130441.1 | 2 | 1107 | ||
| BF591263.1 | 1 | 199 | ||
| X83228.1 | 18 | 3472 | CAA58231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map