Assay Details
Target Gene Details
Entrez Gene ID: | 1429 |
Gene Name: | crystallin zeta |
Gene Aliases: |
- |
Location: |
Chr.1:74705482-74733408 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYZ | NM_001130042.1 | 1 | 130 | NP_001123514.1 |
| NM_001130043.1 | 1 | 130 | NP_001123515.1 | |
| NM_001134759.1 | 1 | 130 | NP_001128231.1 | |
| NM_001889.3 | 1 | 130 | NP_001880.2 | |
| CN368109.1 | 1 | 130 |
Target Gene Details
Entrez Gene ID: | 127253 |
Gene Name: | tRNA-yW synthesizing protein 3 homolog |
Gene Aliases: |
C1orf171 |
Location: |
Chr.1:74733143-74766677 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TYW3 | NM_001162916.1 | 1 | 128 | NP_001156388.1 |
| NM_138467.2 | 1 | 124 | NP_612476.1 | |
| NR_027962.1 | 1 | 102 | ||
| XM_006710347.1 | 1 | 137 | XP_006710410.1 | |
| AK098237.1 | 1 | 124 | BAC05267.1 | |
| AK294264.1 | 1 | 106 | ||
| AK299849.1 | 1 | 79 | ||
| AK311272.1 | 1 | 48 | ||
| BC009514.1 | 1 | 58 | AAH09514.1 | |
| BC071765.1 | 1 | 124 | AAH71765.1 | |
| DA033034.1 | 1 | 128 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830259 | Chr.1:74623493 - 74775378 on Build GRCh38 | Gain |
|
| nsv546579 | Chr.1:74731849 - 74734802 on Build GRCh38 | Loss |
|
| nsv509346 | Chr.1:74724940 - 74739145 on Build GRCh38 | Insertion |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74095164] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map