Assay Details
Target Gene Details
Entrez Gene ID: | 3692 |
Gene Name: | eukaryotic translation initiation factor 6 |
Gene Aliases: |
CAB, EIF3A, ITGB4BP, b(2)gcn, eIF-6, p27(BBP), p27BBP |
Location: |
Chr.20:35278906-35284816 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF6 | NM_001267810.1 | NP_001254739.1 | ||
| NM_002212.3 | NP_002203.1 | |||
| NM_181466.2 | 2 | 155 | NP_852131.1 | |
| NM_181468.2 | NP_852133.1 | |||
| NR_052022.1 | ||||
| NR_052023.1 | ||||
| NR_052024.1 | ||||
| AB062289.1 | BAB93472.1 | |||
| AF022229.1 | AAB97735.1 | |||
| AF047433.1 | AAC39897.1 | |||
| AK296076.1 | ||||
| AK314969.1 | ||||
| AL537456.3 | ||||
| BC001119.2 | AAH01119.1 | |||
| BC011845.2 | AAH11845.1 | |||
| BC019305.1 | AAH19305.1 | |||
| BG283650.1 | ||||
| BM794263.1 | 2 | 109 | ||
| BQ222221.1 | ||||
| CN265282.1 | ||||
| CR456764.1 | CAG33045.1 | |||
| CT002623.1 | ||||
| DB448068.1 | ||||
| Y11435.1 | CAA72243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059168 | Chr.20:35217473 - 35319358 on Build GRCh38 | Loss |
|
| nsv954029 | Chr.20:35284398 - 35298997 on Build GRCh38 | Deletion |
|
| nsv585943 | Chr.20:35247970 - 35297352 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map