Assay Details
Target Gene Details
Entrez Gene ID: | 90527 |
Gene Name: | dual oxidase maturation factor 1 |
Gene Aliases: |
NIP, NUMBIP, mol |
Location: |
Chr.15:45117366-45129938 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
|
| esv1002924 | Chr.15:45120086 - 45132601 on Build GRCh38 | Deletion |
|
| nsv569313 | Chr.15:45121343 - 45165260 on Build GRCh38 | Gain |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map