Assay Details
Target Gene Details
Entrez Gene ID: | 84319 |
Gene Name: | cms1 ribosomal small subunit homolog (yeast) |
Gene Aliases: |
C3orf26 |
Location: |
Chr.3:99817834-100178632 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMSS1 | NM_032359.3 | 1 | 130 | NP_115735.2 |
| AK124387.1 | 1 | 97 | ||
| AK291392.1 | 1 | 102 | ||
| AK312483.1 | 1 | 130 | ||
| BC006475.1 | 1 | 127 | AAH06475.1 | |
| BC006512.2 | 1 | 127 | AAH06512.1 |
Target Gene Details
Entrez Gene ID: | 101929357 |
Gene Name: | uncharacterized LOC101929357 |
Gene Aliases: |
- |
Location: |
Chr.3:99816632-99823865 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HP09053 | NR_110821.1 | 3 | 687 | |
| AB593151.1 | 3 | 687 |
More Information
Set Membership: |
|

Genomic Map