Assay Details
Target Gene Details
Entrez Gene ID: | 5957 |
Gene Name: | recoverin |
Gene Aliases: |
RCV1 |
Location: |
Chr.17:9897710-9905367 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCVRN | NM_002903.2 | 3 | 949 | NP_002894.1 |
| AB593156.1 | 3 | 848 | ||
| BC001720.1 | 3 | 833 | AAH01720.1 | |
| S43855.1 | 3 | 949 | AAB23163.1 | |
| S45545.1 | 3 | 849 | AAB23392.1 | |
| S62028.1 | 3 | 846 | AAB26894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv38n68 | Chr.17:9897032 - 10093580 on Build GRCh38 | Loss |
|
| nsv833360 | Chr.17:9734634 - 9900986 on Build GRCh38 | Loss |
|
| nsv1065401 | Chr.17:9799828 - 9986825 on Build GRCh38 | Loss |
|
| nsv833361 | Chr.17:9785863 - 9979090 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map