Assay Details
Target Gene Details
Entrez Gene ID: | 57124 |
Gene Name: | CD248 molecule |
Gene Aliases: |
CD164L1, TEM1 |
Location: |
Chr.11:66314487-66317044 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD248 | NM_020404.2 | 1 | 2050 | NP_065137.1 |
| AF279142.1 | 1 | 2038 | AAG00867.1 | |
| AJ295846.1 | 1 | 2050 | CAC34381.1 | |
| AK027290.1 | 1 | 1172 | BAB55018.1 | |
| BC051340.1 | 1 | 2037 | AAH51340.1 | |
| BC104484.1 | 1 | 2050 | AAI04485.1 | |
| BC105633.1 | 1 | 2050 | AAI05634.1 | |
| BQ005949.1 | 1 | 527 | ||
| KU215671.1 | ||||
| KU257670.1 |
More Information
Set Membership: |
|

Genomic Map