Assay Details
Target Gene Details
Entrez Gene ID: | 9755 |
Gene Name: | TBK1 binding protein 1 |
Gene Aliases: |
ProSAPiP2, SINTBAD |
Location: |
Chr.17:47694065-47712063 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBKBP1 | NM_014726.2 | 1 | 990 | NP_055541.1 |
| XM_005257859.4 | 2 | 289 | XP_005257916.1 | |
| XM_005257860.4 | 2 | 1114 | XP_005257917.1 | |
| XM_005257861.4 | 2 | 594 | XP_005257918.1 | |
| XM_005257862.3 | 2 | 214 | XP_005257919.1 | |
| XM_017025431.1 | 1 | 1498 | XP_016880920.1 | |
| XM_017025432.1 | 1 | 1500 | XP_016880921.1 | |
| AB018318.1 | 1 | 990 | ||
| BC111418.1 | 1 | 569 | ||
| BC152407.1 | 1 | 990 | ||
| BC167150.1 | 1 | 990 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9570 | Chr.17:47693507 - 47702425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map