Assay Details
Target Gene Details
Entrez Gene ID: | 22900 |
Gene Name: | caspase recruitment domain family member 8 |
Gene Aliases: |
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN |
Location: |
Chr.19:48203148-48256269 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3613n100 | Chr.19:48220413 - 48256972 on Build GRCh38 | Gain |
|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss |
|
| nsv469623 | Chr.19:48209728 - 48360236 on Build GRCh38 | Loss |
|
| nsv458716 | Chr.19:48208219 - 48239130 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map