Assay Details
Target Gene Details
Entrez Gene ID: | 1580 |
Gene Name: | cytochrome P450 family 4 subfamily B member 1 |
Gene Aliases: |
CYPIVB1, P-450HP |
Location: |
Chr.1:46798995-46819349 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4B1 | NM_000779.3 | 5 | 641 | NP_000770.2 |
| NM_001099772.1 | 5 | 641 | NP_001093242.1 | |
| NM_001319161.1 | 4 | 596 | NP_001306090.1 | |
| NM_001319162.1 | 3 | 454 | NP_001306091.1 | |
| NM_001319163.1 | 3 | 454 | NP_001306092.1 | |
| NR_135003.1 | 5 | 641 | ||
| XM_011540832.1 | 5 | 644 | XP_011539134.1 | |
| XM_011540833.1 | 4 | 276 | XP_011539135.1 | |
| XM_017000466.1 | 4 | 548 | XP_016855955.1 | |
| AF491285.1 | 5 | 557 | AAM09532.1 | |
| AK225576.1 | 5 | 593 | ||
| AK225578.1 | 5 | 593 | ||
| AK300921.1 | 3 | 454 | ||
| AK300941.1 | 3 | 303 | ||
| AK307715.1 | 5 | 715 | ||
| AK313170.1 | 5 | 593 | ||
| AY064484.1 | 5 | 557 | AAL57719.1 | |
| AY064485.1 | 5 | 557 | AAL57720.1 | |
| AY064486.1 | 5 | 557 | AAL57721.1 | |
| AY151048.1 | 5 | 557 | AAN72311.1 | |
| AY151049.1 | 4 | 512 | AAN72312.1 | |
| BC017758.1 | 5 | 593 | AAH17758.1 | |
| CD013981.1 | 5 | 575 | ||
| DC358497.1 | 3 | 406 | ||
| J02871.1 | 5 | 569 | AAA35712.1 | |
| X16699.1 | 5 | 557 | CAA34672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585939 | Chr.1:46798765 - 46837514 on Build GRCh38 | Gain |
|
| esv3585938 | Chr.1:46730796 - 46989113 on Build GRCh38 | Gain |
|
| nsv1011086 | Chr.1:46793406 - 46839519 on Build GRCh38 | Gain |
|
| nsv1010063 | Chr.1:46798533 - 46819446 on Build GRCh38 | Gain |
|
| dgv160n100 | Chr.1:46805270 - 46856310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map